2,6-Bis(4-fluorobenzylidene)cyclohexanone ≥96.0% (by HPLC)

供应商: TCI

Synonyms: 2,6-Bis(4-fluorobenzal)cyclohexanone

B2282-25G B2282-5G
TCIAB2282-25GEA 0 CNY
TCIAB2282-25G TCIAB2282-5G
2,6-Bis(4-fluorobenzylidene)cyclohexanone ≥96.0% (by HPLC)
2,6-Bis(4-fluorobenzylidene)cyclohexanone
分子式: C₂₀H₁₆F₂O
MDL编号: MFCD00112608
CAS号: 62085-74-9

现在订购

产品技术规格

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 96.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR