Boc-D-Ser-OH ≥98.0% (通过HPLC,滴定分析)

供应商: TCI

B2258-1G B2258-5G
TCIAB2258-1GEA 0 CNY
TCIAB2258-1G TCIAB2258-5G
Boc-D-Ser-OH ≥98.0% (通过HPLC,滴定分析)
Boc-D-Ser-OH
分子式: C₈H₁₅NO₅
MDL编号: MFCD00063142
CAS号: 6368-20-3

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +7.0 to +10.0 deg (C=2, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR