N-叔丁基苯亚磺酰胺 ≥96.0% (by GC)

供应商: TCI

Synonyms: N-(1,1-二甲基乙基)苯亚磺酰胺, N-叔丁基苯磺酰胺

B2240-1G B2240-25G B2240-5G
TCIAB2240-1GEA 0 CNY
TCIAB2240-1G TCIAB2240-25G TCIAB2240-5G
N-叔丁基苯亚磺酰胺 ≥96.0% (by GC)
N-叔丁基苯亚磺酰胺
分子式: C₁₀H₁₅NS
存储条件: Refrigerator
MDL编号: MFCD03844776
CAS号: 19117-31-8

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR