N-Benzylidenebenzenesulphonamide ≥98.0% (by HPLC, total nitrogen)
供应商: TCI
|
Warning
|
B2206-25G
B2206-5G
TCIAB2206-25GEA
0
CNY
TCIAB2206-25G
TCIAB2206-5G
N-Benzylidenebenzenesulphonamide ≥98.0% (by HPLC, total nitrogen)
N-Benzylidenebenzenesulphonamide
分子式:
C₁₃H₁₁NO₂S |
MDL编号:
MFCD00011587 CAS号: 13909-34-7 |
产品技术规格
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 80.0 to 86.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...