N-Bromosaccharin ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 2-Bromo-1,2-benzisothiazol-3(2H)-one 1,1-Dioxide, 2-Bromobenzo[d]isothiazol-3(2H)-one 1,1-dioxide

B2152-25G B2152-5G
TCIAB2152-25GEA 0 CNY
TCIAB2152-25G TCIAB2152-5G
N-Bromosaccharin ≥98.0% (by titrimetric analysis)
N-Bromosaccharin
分子式: C₇H₄BrNO₃S
分子量: 262.08 g/mol
存储条件: Refrigerator
MDL编号: MFCD03844772
CAS号: 35812-01-2
EINECS: 252-737-4

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR