2-(4-Bromobutyl)-1,3-dioxolane ≥95.0% (by GC)

供应商: TCI

Synonyms: 5-Bromovaleraldehyde Ethylene Acetal

B2128-25G B2128-5G
TCIAB2128-25GEA 0 CNY
TCIAB2128-25G TCIAB2128-5G
2-(4-Bromobutyl)-1,3-dioxolane ≥95.0% (by GC)
2-(4-Bromobutyl)-1,3-dioxolane
分子式: C₇H₁₃BrO₂
分子量: 209.08 g/mol
存储条件: Refrigerator
MDL编号: MFCD02093454
CAS号: 87227-41-6

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR