(S)-1-Boc-2-azetidinemethanol ≥95.0% (by GC)
供应商: TCI
Synonyms:
(S)-1-Boc-2-Azetidinemethanol, (S)-1-Boc-2-azetidinemethanol, (S)-1-(tert-Butoxycarbonyl)-2-azetidinemethanol
B2123-1G
B2123-100MG
TCIAB2123-1GEA
0
CNY
TCIAB2123-1G
TCIAB2123-100MG
(S)-1-Boc-2-azetidinemethanol ≥95.0% (by GC)
(S)-1-Boc-2-azetidinemethanol
分子式:
C₉H₁₇NO₃ 分子量: 187.2362 g/mol |
MDL编号:
MFCD03093620 CAS号: 161511-85-9 |
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Optical purity (LC) | min. 98.0 ee% |
Specific rotation [a]20/D | -72.0 to -76.0 deg (C=1, MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...