3,5-Bis[3,5-bis(benzyloxy)benzyloxy]benzyl bromide ≥97.0% (by HPLC)

供应商: TCI
Warning

Synonyms: 3,5-Bis(3,5-dibenzyloxybenzyloxy)benzyl Bromide

B2118-1G B2118-5G
TCIAB2118-1GEA 0 CNY
TCIAB2118-1G TCIAB2118-5G
3,5-Bis[3,5-bis(benzyloxy)benzyloxy]benzyl bromide ≥97.0% (by HPLC)
3,5-Bis[3,5-bis(benzyloxy)benzyloxy]benzyl bromide
分子式: C₄₉H₄₃BrO₆
分子量: 807.78 g/mol
存储条件: Refrigerator
MDL编号: MFCD02093449
CAS号: 129536-41-0

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Melting point 127.0 to 131.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR