(1S,2R)-2-[N-Benzyl-N-(mesitylenesulphonyl)amino]-1-phenyl-1-propanol ≥98.0% (by HPLC)

供应商: TCI

Synonyms: (1S,2R)-N-Benzyl-N-(mesitylenesulphonyl)norephedrine, (1S,2R)-2-[N-Benzyl-N-(2,4,6-trimethylbenzenesulphonyl)amino]-1-phenyl-1-propanol

B2104-1G B2104-5G
TCIAB2104-1GEA 0 CNY
TCIAB2104-1G TCIAB2104-5G
(1S,2R)-2-[N-Benzyl-N-(mesitylenesulphonyl)amino]-1-phenyl-1-propanol ≥98.0% (by HPLC)
(1S,2R)-2-[N-Benzyl-N-(mesitylenesulphonyl)amino]-1-phenyl-1-propanol
分子式: C₂₅H₂₉NO₃S
分子量: 423.57 g/mol
MDL编号: MFCD02093447
CAS号: 187324-64-7

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 98.0 ee%
Melting point 130.0 to 134.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR