(4S,5S)-2,2-二甲基--1,3-二氧戊环-4,5-二基)双(二苯基甲醇) ≥97.0% (by HPLC)
供应商: TCI
Synonyms:
(4S,5S)-4,5-双(二苯基羟甲基)-2,2-二甲基二氧戊环, (+)-2,3-O-异亚丙基-1,1,4,4-四苯基-D-苏糖醇, (+)-反式-α,α′-(2,2-二甲基-1,3-二氧戊环-4,5-二基)双(二苯基甲醇)
B2048-1G
B2048-5G
TCIAB2048-1GEA
0
CNY
TCIAB2048-1G
TCIAB2048-5G
(4S,5S)-2,2-二甲基--1,3-二氧戊环-4,5-二基)双(二苯基甲醇) ≥97.0% (by HPLC)
(4S,5S)-2,2-二甲基--1,3-二氧戊环-4,5-二基)双(二苯基甲醇)
分子式:
C₃₁H₃₀O₄ 分子量: 466.58 g/mol |
MDL编号:
MFCD00010079 CAS号: 93379-49-8 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Optical purity (LC) | min. 98.0 ee% |
Melting point | 194.0 to 198.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...