1-叔丁氧基羰基-1,2,4-三唑 ≥97.0% (by GC)

供应商: TCI

Synonyms: 1-叔丁氧基羰基-1H-1,2,4-三唑

B1969-25G B1969-5G
TCIAB1969-25GEA 0 CNY
TCIAB1969-25G TCIAB1969-5G
1-叔丁氧基羰基-1,2,4-三唑 ≥97.0% (by GC)
1-叔丁氧基羰基-1,2,4-三唑
分子式: C₇H₁₁N₃O₂
分子量: 169.18 g/mol
存储条件: Refrigerator
MDL编号: MFCD00042665
CAS号: 41864-24-8
EINECS: 255-570-5

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 44.0 to 48.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR