4-Butylcyclohexanol (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (通过GC)
供应商: TCI
B1930-25G
B1930-5G
TCIAB1930-25GEA
0
CNY
TCIAB1930-25G
TCIAB1930-5G
4-Butylcyclohexanol (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (通过GC)
4-Butylcyclohexanol (cis- and trans- mixture)
分子式:
C₁₀H₂₀O 沸点: 122°C |
MDL编号:
MFCD01321147 CAS号: 70568-60-4 |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % (total of isomer) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...