N-Benzoyl-L-tyrosine ≥98.0% (by HPLC, titration analysis)

供应商: TCI

B1911-25G B1911-5G
TCIAB1911-25GEA 0 CNY
TCIAB1911-25G TCIAB1911-5G
N-Benzoyl-L-tyrosine ≥98.0% (by HPLC, titration analysis)
N-Benzoyl-L-tyrosine
分子式: C₁₆H₁₅NO₄
分子量: 285.30 g/mol
MDL编号: MFCD01321140
CAS号: 2566-23-6

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 163.0 to 167.0 deg-C
Specific rotation [a]20/D -75.0 to -80.0 deg (C=1, DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR