2,2′-联噻吩-5-甲醛 ≥98.0% (by GC)
供应商: TCI
Synonyms:
2,2-Bithiophene-5-carboxaldehyde, 5-Formyl-2,2'-bithiophene
B1838-1G
B1838-25G
B1838-5G
TCIAB1838-1GEA
0
CNY
TCIAB1838-1G
TCIAB1838-25G
TCIAB1838-5G
2,2′-联噻吩-5-甲醛 ≥98.0% (by GC)
2,2′-联噻吩-5-甲醛
分子式:
C₉H₆OS₂ 分子量: 194.27 g/mol 沸点: 210°C |
MDL编号:
MFCD00052294 CAS号: 3779-27-9 |
产品技术规格
Appearance | White to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 55.0 to 58.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...