双酚 A 二缩水甘油醚 ≥85.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
2,2-二[4-(氧化缩水甘油)苯基]丙烷, D.E.R. 332, BADGE, 4,4′-异亚丙基联苯酚二缩水甘油醚
B1796-25G
B1796-500G
TCIAB1796-25GEA
0
CNY
TCIAB1796-25G
TCIAB1796-500G
双酚 A 二缩水甘油醚 ≥85.0% (by GC)
双酚 A 二缩水甘油醚
分子式:
C₂₁H₂₄O₄ 分子量: 340.42 g/mol 沸点: 210°C |
MDL编号:
MFCD00080480 CAS号: 1675-54-3 EINECS: 216-823-5 |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 85.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...