双酚芴 ≥96.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
9,9-二(4-羟苯基)芴
B1715-25G
B1715-500G
TCIAB1715-25GEA
0
CNY
TCIAB1715-25G
TCIAB1715-500G
双酚芴 ≥96.0% (by GC)
双酚芴
分子式:
C₂₅H₁₈O₂ |
MDL编号:
MFCD00191392 CAS号: 3236-71-3 EINECS: 406-950-6 UN号: 3077 ADR: 9,III |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 224.0 to 228.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...