N-叔丁基-α-苯基硝酮 ≥98.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: 苯基·N-叔丁基硝酮, PBN, N-苯亚甲基-叔丁胺-N-氧化物

B1701-1G B1701-5G
TCIAB1701-1GEA 0 CNY
TCIAB1701-1G TCIAB1701-5G
N-叔丁基-α-苯基硝酮 ≥98.0% (by HPLC, titration analysis)
N-叔丁基-α-苯基硝酮
分子式: C₁₁H₁₅NO
MDL编号: MFCD00008799
CAS号: 3376-24-7
EINECS: 222-168-6

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 72.0 to 75.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR