1,2-双(氯二甲基硅基)乙烷 ≥95.0% (by GC) protecting reagent for primary amines

供应商: TCI
Danger

Synonyms: 2,5-二氯-2,5-二甲基-2,5-二硅杂己烷, 1,1,4,4-四甲基-1,4-二氯乙烯乙硅烷

B1688-25G B1688-5G
TCIAB1688-25GEA 0 CNY
TCIAB1688-25G TCIAB1688-5G
1,2-双(氯二甲基硅基)乙烷 ≥95.0% (by GC) protecting reagent for primary amines
1,2-双(氯二甲基硅基)乙烷
分子式: C₆H₁₆Cl₂Si₂
分子量: 215.26 g/mol
闪点: 65°C
MDL编号: MFCD00000505
CAS号: 13528-93-3
EINECS: 236-871-0
UN号: 2987
ADR: 8 II

现在订购

产品技术规格

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR