2,2-Bis(4-oxocyclohexyl)propane ≥95.0% (by GC)
供应商: TCI
Synonyms:
4,4'-Isopropylidenebis(cyclohexanone)
B1672-25G
B1672-5G
TCIAB1672-25GEA
0
CNY
TCIAB1672-25G
TCIAB1672-5G
2,2-Bis(4-oxocyclohexyl)propane ≥95.0% (by GC)
2,2-Bis(4-oxocyclohexyl)propane
分子式:
C₁₅H₂₄O₂ 分子量: 236.36 g/mol |
MDL编号:
MFCD00191618 CAS号: 7418-16-8 EINECS: 231-035-1 |
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 161.0 to 165.0 deg-C |
Solubility in Chloroform | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...