1,1-Bis(methylthio)ethylene ≥90.0% (by GC)
供应商: TCI
Synonyms:
Ketene Dimethyl Thioacetal
B1650-1G
B1650-5G
TCIAB1650-1GEA
0
CNY
TCIAB1650-1G
TCIAB1650-5G
1,1-Bis(methylthio)ethylene ≥90.0% (by GC)
1,1-Bis(methylthio)ethylene
分子式:
C₄H₈S₂ 沸点: 110°C 存储条件: Refrigerator |
MDL编号:
MFCD00191617 CAS号: 51102-74-0 |
产品技术规格
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...