Boc-Glu(OBzl)-OH ≥98.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: Boc-L-谷氨酸-5-苄酯

B1633-25G B1633-5G
TCIAB1633-25GEA 0 CNY
TCIAB1633-25G TCIAB1633-5G
Boc-Glu(OBzl)-OH ≥98.0% (by HPLC, titration analysis)
Boc-Glu(OBzl)-OH
分子式: C₁₇H₂₃NO₆
分子量: 337.37 g/mol
MDL编号: MFCD00065569
CAS号: 13574-13-5
EINECS: 237-007-5

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 66.0 to 71.0 deg-C
Specific rotation [a]20/D -16.0 to -17.0 deg (C=2, DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR