Boc-Tyr(Bzl)-OH ≥98.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: Boc-O-苄基-L-酪氨酸

B1631-25G B1631-5G
TCIAB1631-25GEA 0 CNY
TCIAB1631-25G TCIAB1631-5G
Boc-Tyr(Bzl)-OH ≥98.0% (by HPLC, titration analysis)
Boc-Tyr(Bzl)-OH
分子式: C₂₁H₂₅NO₅
分子量: 371.43 g/mol
MDL编号: MFCD00065597
CAS号: 2130-96-3
EINECS: 218-349-4

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 109.0 to 113.0 deg-C
Specific rotation [a]20/D +25.0 to +30.0 deg (C=2, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR