癸酸乙酯 99+%

供应商: Thermo Fisher Scientific

Synonyms: 羊蜡酸乙酯, 正癸酸乙酯

165000050 165001000 165005000
ACRO165000050EA 0 CNY
ACRO165000050 ACRO165001000 ACRO165005000
癸酸乙酯 99+%
癸酸乙酯
分子式: C₁₂H₂₄O₂
分子量: 200.32 g/mol
沸点: 245 °C (760 mmHg)
溶点: –20 °C
密度: 0.8665 g/cm³
闪点: 102 °C
存储条件: Ambient
MDL编号: MFCD00009581
CAS号: 110-38-3
EINECS: 203-761-9
Merck Index: 13,03811

现在订购

产品技术规格

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4240 to 1.4270 (20°C, 589 nm)
Water ≤0.3 % (K.F.)
Density (20°C) 0.864 to 0.869 kg/l
GC ≥99.0 %
Acid value ≤1 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR