1-Bromo-4-decylbenzene ≥88.0% (by GC)

供应商: TCI

Synonyms: 4-Decylbromobenzene

B1611-1G B1611-25G B1611-5G
TCIAB1611-1GEA 0 CNY
TCIAB1611-1G TCIAB1611-25G TCIAB1611-5G
1-Bromo-4-decylbenzene ≥88.0% (by GC)
1-Bromo-4-decylbenzene
分子式: C₁₆H₂₅Br
分子量: 297.28 g/mol
MDL编号: MFCD00061112
CAS号: 106418-67-1

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 88.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR