7-Bromobenz[a]anthracene ≥98.0% (by GC)

供应商: TCI

B1603-1G B1603-5G
TCIAB1603-1GEA 0 CNY
TCIAB1603-1G TCIAB1603-5G
7-Bromobenz[a]anthracene ≥98.0% (by GC)
7-Bromobenz[a]anthracene
分子式: C₁₈H₁₁Br
分子量: 307.19 g/mol
MDL编号: MFCD00029016
CAS号: 32795-84-9

现在订购

产品技术规格

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 151.0 to 154.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR