2-Bromo-1,4-benzoquinone ≥93.0% (by GC)

供应商: TCI
Warning

Synonyms: 2-Bromocyclohexadiene-1,4-dione, Bromoquinone

B1601-1G B1601-5G
TCIAB1601-1GEA 0 CNY
TCIAB1601-1G TCIAB1601-5G
2-Bromo-1,4-benzoquinone ≥93.0% (by GC)
2-Bromo-1,4-benzoquinone
分子式: C₆H₃BrO₂
存储条件: Refrigerator
MDL编号: MFCD00189377
CAS号: 3958-82-5

现在订购

产品技术规格

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 93.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR