2-(2-Bromoethyl)-2-methyl-1,3-dioxolane ≥96.0% (by GC)
供应商: TCI
|
Warning
|
B1598-25G
B1598-5G
TCIAB1598-25GEA
0
CNY
TCIAB1598-25G
TCIAB1598-5G
2-(2-Bromoethyl)-2-methyl-1,3-dioxolane ≥96.0% (by GC)
2-(2-Bromoethyl)-2-methyl-1,3-dioxolane
分子式:
C₆H₁₁BrO₂ 分子量: 195.06 g/mol 沸点: 74°C 存储条件: Refrigerator |
MDL编号:
MFCD00191321 CAS号: 37865-96-6 |
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...