(3S)-(+)-1-Benzyl-3-(methylamino)pyrrolidine ≥98.0% (by GC)

供应商: TCI
Danger

B1583-25G B1583-5G
TCIAB1583-25GEA 0 CNY
TCIAB1583-25G TCIAB1583-5G
(3S)-(+)-1-Benzyl-3-(methylamino)pyrrolidine ≥98.0% (by GC)
(3S)-(+)-1-Benzyl-3-(methylamino)pyrrolidine
分子式: C₁₂H₁₈N₂
分子量: 190.29 g/mol
MDL编号: MFCD00191310
CAS号: 169749-99-9
UN号: 2735
ADR: 8 III

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 98.0 ee%
Specific rotation [a]20/D +2.8 to +3.2 deg (C=10, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR