cis-1,2-Dicyano-1,2-bis(2,4,5-trimethyl-3-thienyl)ethene ≥98.0% (by total nitrogen basis)

供应商: TCI
Danger

Synonyms: 1,2-Bis(2,4,5-trimethyl-3-thienyl)-cis-1,2-dicyanoethene, (Z)-2,3-Bis(2,4,5-trimethylthiophen-3-yl)maleonitrile

B1536-1G B1536-5G
TCIAB1536-1GEA 0 CNY
TCIAB1536-1G TCIAB1536-5G
cis-1,2-Dicyano-1,2-bis(2,4,5-trimethyl-3-thienyl)ethene ≥98.0% (by total nitrogen basis)
cis-1,2-Dicyano-1,2-bis(2,4,5-trimethyl-3-thienyl)ethene
分子式: C₁₈H₁₈N₂S₂
分子量: 326.48 g/mol
MDL编号: MFCD00143977
CAS号: 112440-46-7
UN号: 3439
ADR: 6.1 III

现在订购

产品技术规格

Appearance Light yellow to Brown powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 120.0 to 124.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR