Behenic anhydride ≥97.0% (by GC)

供应商: TCI
Warning

Synonyms: Docosanoic Anhydride

B1531-1G B1531-5G
TCIAB1531-1GEA 0 CNY
TCIAB1531-1G TCIAB1531-5G
Behenic anhydride ≥97.0% (by GC)
Behenic anhydride
分子式: C₄₄H₈₆O₃
MDL编号: MFCD00010452
CAS号: 55726-23-3

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 81.0 to 85.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR