叔丁基 2,2,2-三氯乙酰亚胺酯 ≥95.0% (by GC)

供应商: TCI
Warning

Synonyms: TBTA

B1496-25ML B1496-5ML
TCIAB1496-25MLEA 0 CNY
TCIAB1496-25ML TCIAB1496-5ML
叔丁基 2,2,2-三氯乙酰亚胺酯 ≥95.0% (by GC)
叔丁基 2,2,2-三氯乙酰亚胺酯
分子式: C₆H₁₀Cl₃NO
分子量: 218.502 g/mol
沸点: 69°C
闪点: 55°C
存储条件: Refrigerator
MDL编号: MFCD00077410
CAS号: 98946-18-0
UN号: 1993
ADR: 3 III

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR