羧乙基锗倍半氧化物 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: GE 132

B1367-25G B1367-5G
TCIAB1367-25GEA 0 CNY
TCIAB1367-25G TCIAB1367-5G
羧乙基锗倍半氧化物 ≥98.0% (by titrimetric analysis)
羧乙基锗倍半氧化物
分子式: C₆H₁₀Ge₂O₇
分子量: 339.40 g/mol
MDL编号: MFCD00049319
CAS号: 12758-40-6
EINECS: 235-800-0
Merck Index: 14,07795

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR