Bis(2-ethoxyethyl)amine ≥98.0% (通过GC)
供应商: TCI
|
Danger
|
B1364-25ML
B1364-500ML
TCIAB1364-25MLEA
0
CNY
TCIAB1364-25ML
TCIAB1364-500ML
Bis(2-ethoxyethyl)amine ≥98.0% (通过GC)
Bis(2-ethoxyethyl)amine
分子式:
C₈H₁₉NO₂ 沸点: 200°C |
MDL编号:
MFCD00039904 CAS号: 124-21-0 UN号: 2735 ADR: 8,III |
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...