N,N′-(4,4′-亚甲基二苯基)双马来酰亚胺 ≥96.0% (by GC)

供应商: TCI
Danger

Synonyms: 双马来酰亚胺

B1109-25G B1109-500G
TCIAB1109-25GEA 0 CNY
TCIAB1109-25G TCIAB1109-500G
N,N′-(4,4′-亚甲基二苯基)双马来酰亚胺 ≥96.0% (by GC)
N,N′-(4,4′-亚甲基二苯基)双马来酰亚胺
分子式: C₂₁H₁₄N₂O₄
分子量: 358.35 g/mol
MDL编号: MFCD00005507
CAS号: 13676-54-5
EINECS: 237-163-4
UN号: 2811
ADR: 6.1 II

现在订购

产品技术规格

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 96.0 %
Melting point 157.0 to 160.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR