2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol ≥98.0% (通过HPLC,滴定分析)

供应商: TCI
Warning

B1081-1G B1081-100MG
TCIAB1081-1GEA 0 CNY
TCIAB1081-1G TCIAB1081-100MG
2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol ≥98.0% (通过HPLC,滴定分析)
2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol
分子式: C₁₅H₁₇BrN₄O
MDL编号: MFCD00006255
CAS号: 14337-53-2
EINECS: 238-286-6

现在订购

产品技术规格

Appearance Orange to Brown to Dark red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR