2-(叔丁氧羰基氧亚氨基)-2-苯乙腈 ≥98.0% (by total nitrogen basis)

供应商: TCI
Danger

Synonyms: Boc-ON, 2-(tert-Butoxycarbonyloxyimino)-2-phenyl­acetonitrile

B0988-25G B0988-5G
TCIAB0988-25GEA 0 CNY
TCIAB0988-25G TCIAB0988-5G
2-(叔丁氧羰基氧亚氨基)-2-苯乙腈 ≥98.0% (by total nitrogen basis)
2-(叔丁氧羰基氧亚氨基)-2-苯乙腈
分子式: C₁₃H₁₄N₂O₃
分子量: 246.27 g/mol
存储条件: Refrigerator
MDL编号: MFCD00001863
CAS号: 58632-95-4
EINECS: 261-370-9
UN号: 3439
ADR: 6.1 III

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 94.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 85.0 to 89.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR