Butylamine hydrochloride ≥98.0% (通过滴定分析)

供应商: TCI
Warning

B0710-25G B0710-500G
TCIAB0710-25GEA 0 CNY
TCIAB0710-25G TCIAB0710-500G
Butylamine hydrochloride ≥98.0% (通过滴定分析)
Butylamine hydrochloride
分子式: C₄H₁₁N
MDL编号: MFCD00043791
CAS号: 3858-78-4
EINECS: 223-369-1

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Melting point 212.0 to 217.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR