Tri-o-tolyl borate ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: Boric Acid Tri-o-cresyl Ester, Boric Acid Tri-o-tolyl Ester, Tri-o-cresyl Borate

B0519-25ML B0519-5ML
TCIAB0519-25MLEA 0 CNY
TCIAB0519-25ML TCIAB0519-5ML
Tri-o-tolyl borate ≥98.0% (by titrimetric analysis)
Tri-o-tolyl borate
分子式: C₂₁H₂₁BO₃
分子量: 332.21 g/mol
沸点: 386°C
闪点: 174°C
MDL编号: MFCD00059311
CAS号: 2665-12-5

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR