Leuco malachite green ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 4,4'-Bis(dimethylamino)triphenylmethane, 4,4'-(Phenylmethylene)bis(N,N-dimethylaniline), 4,4'-Tetramethyldiaminotriphenylmethane, 4,4'-Benzylidenebis(N,N-dimethylaniline), Leucomalachite green

B0419-1G B0419-25G
TCIAB0419-1GEA 0 CNY
TCIAB0419-1G TCIAB0419-25G
Leuco malachite green ≥98.0% (by titrimetric analysis)
Leuco malachite green
分子式: C₂₃H₂₆N₂
分子量: 330.48 g/mol
MDL编号: MFCD00008315
CAS号: 129-73-7
EINECS: 204-961-9

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 100.0 to 103.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR