Benzoylurea ≥98.0% (通过HPLC,总氮)
供应商: TCI
|
Warning
|
B0400-25G
B0400-500G
TCIAB0400-25GEA
0
CNY
TCIAB0400-25G
TCIAB0400-500G
Benzoylurea ≥98.0% (通过HPLC,总氮)
Benzoylurea
分子式:
C₈H₈N₂O₂ |
MDL编号:
MFCD00025441 CAS号: 614-22-2 EINECS: 210-372-8 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 208.0 to 220.0 deg-C |
Solubility in N,N-DMF | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...