2,3-Diphenyl-5-(2-thienyl)tetrazolium chloride ≥97.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: STC

B0325-1G B0325-100MG
TCIAB0325-1GEA 0 CNY
TCIAB0325-1G TCIAB0325-100MG
2,3-Diphenyl-5-(2-thienyl)tetrazolium chloride ≥97.0% (by HPLC, titration analysis)
2,3-Diphenyl-5-(2-thienyl)tetrazolium chloride
分子式: C₁₇H₁₃ClN₄S
MDL编号: MFCD00060088
CAS号: 38800-20-3

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR