2,4,5-Trichlorobenzenesulphonic acid hydrate ≥98.0% (通过滴定分析)
供应商: TCI
|
Danger
|
B0248-25G
B0248-5G
TCIAB0248-25GEA
0
CNY
TCIAB0248-25G
TCIAB0248-5G
2,4,5-Trichlorobenzenesulphonic acid hydrate ≥98.0% (通过滴定分析)
2,4,5-Trichlorobenzenesulphonic acid hydrate
分子式:
C₆H₃Cl₃O₃S |
MDL编号:
MFCD00070539 CAS号: 6378-25-2 EINECS: 228-950-3 UN号: 2585 ADR: 8,III |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % (calcd.on anh.substance) |
Solubility in Water | almost transparency |
Water | 6.0 to 8.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...