Benzoyl-L-histidine monohydrate ≥98.0% (通过HPLC,滴定分析)
供应商: TCI
B0204-1G
B0204-100MG
TCIAB0204-1GEA
0
CNY
TCIAB0204-1G
TCIAB0204-100MG
Benzoyl-L-histidine monohydrate ≥98.0% (通过HPLC,滴定分析)
Benzoyl-L-histidine monohydrate
分子式:
C₁₃H₁₃N₃O₃ |
MDL编号:
MFCD00037850 CAS号: 5354-94-9 EINECS: 226-332-8 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % (calcd.on anh.substance) |
Specific rotation [a]20/D | -46.0 to -48.0 deg (C=2, 0.5mol/L HCl) |
Solubility in 0.5mol/L Hydrochloric acid | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...