Benzal diacetate ≥95.0% (通过GC)
供应商: TCI
B0010-25G
B0010-500G
TCIAB0010-25GEA
0
CNY
TCIAB0010-25G
TCIAB0010-500G
Benzal diacetate ≥95.0% (通过GC)
Benzal diacetate
分子式:
C₁₁H₁₂O₄ 沸点: 154°C |
MDL编号:
MFCD00059206 CAS号: 581-55-5 EINECS: 209-469-8 |
产品技术规格
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 95.0 % |
Melting point | 42.0 to 46.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...