3-Bromomethyl-7-methoxy-1,4-benzoxazin-2-one ≥98.0% (by titrimetric analysis) for HPLC labeling
供应商: TCI
A5553-1G
A5553-100MG
TCIAA5553-1GEA
0
CNY
TCIAA5553-1G
TCIAA5553-100MG
3-Bromomethyl-7-methoxy-1,4-benzoxazin-2-one ≥98.0% (by titrimetric analysis) for HPLC labeling
3-Bromomethyl-7-methoxy-1,4-benzoxazin-2-one
分子式:
C₁₀H₈BrNO₃ 分子量: 270.08 g/mol 存储条件: Refrigerator |
MDL编号:
MFCD00143099 CAS号: 124522-09-4 |
产品技术规格
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Argentmetric Titration) | min. 98.0 % |
Effect test of HPLC derivatization | Effective as labeling agent for Linoleic Acid , Oleic Acid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...