2,3,4,6-四-O-苯甲酰基-β-D-吡喃葡萄糖基异硫氰酸酯 ≥98.0% (by HPLC) for HPLC labeling

供应商: TCI
Warning

Synonyms: BGITC

A5515-1G A5515-100MG
TCIAA5515-1GEA 0 CNY
TCIAA5515-1G TCIAA5515-100MG
2,3,4,6-四-O-苯甲酰基-β-D-吡喃葡萄糖基异硫氰酸酯 ≥98.0% (by HPLC) for HPLC labeling
2,3,4,6-四-O-苯甲酰基-β-D-吡喃葡萄糖基异硫氰酸酯
分子式: C₃₅H₂₇NO₉S
分子量: 637.66 g/mol
MDL编号: MFCD00075064
CAS号: 132413-50-4

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 149.0 to 153.0 deg-C
Effect test of HPLC derivatization Effective as labeling agent for DL-Alanine.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR