Xylylazo violet I spectrophotometric reagent for Mg

供应商: TCI

A5007-1G A5007-5G
TCIAA5007-1GEA 0 CNY
TCIAA5007-1G TCIAA5007-5G
Xylylazo violet I spectrophotometric reagent for Mg
Xylylazo Violet I
分子式: C₂₅H₂₁N₃O₆S
MDL编号: MFCD00003890
CAS号: 14936-97-1
EINECS: 239-012-8

现在订购

产品技术规格

Appearance Dark red to Brown powder to crystal
Absorbance min. 0.450[14mg/L, Borate buffer+EtOH (50 vol%), 606.5 nm] (calcd.on dried substance)
Drying loss max. 7.0 %
Suitability for Magnesium-analysis Abs min. 0.700 (14mg/L , near 504nn) in the presence of Magnesium


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR