Xylylazo violet II ≥95.0% (by HPLC) spectrophotometric reagent for Mg

供应商: TCI

Synonyms: Xylidyl Blue II, 1-Azo-2-hydroxy-3-(2,4-dimethylcarboxyanilido)naphthalene-1-(2-hydroxybenzene), 2-[2-Hydroxy-3-(2,4-xylylcarbamoyl)-1-naphthylazo]phenol

A5006-1G A5006-5G
TCIAA5006-1GEA 0 CNY
TCIAA5006-1G TCIAA5006-5G
Xylylazo violet II ≥95.0% (by HPLC) spectrophotometric reagent for Mg
Xylylazo violet II
分子式: C₂₅H₂₁N₃O₃
分子量: 411.46 g/mol
溶点: 247°C
MDL编号: MFCD00014309
CAS号: 523-67-1
EINECS: 208-347-1

现在订购

产品技术规格

Appearance Orange to Amber to Dark red powder to crystal
Purity (HPLC) min. 95.0 area%
Suitability for Magnesium-analysis Abs min. 0.300 (40mg/L , near 505nn) in the presence of Magnesium


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR