Alendronate sodium trihydrate ≥97.0% (by titrimetric analysis)

供应商: TCI

Synonyms: (4-Amino-1-hydroxy-1-phosphonobutyl)phosphonic Acid Monosodium Salt, Sodium hydrogen (4-amino-1-hydroxy-1-phosphonobutyl)phosphonate trihydrate, Alendronic Acid Monosodium Salt, Alendronate sodium hydrate, Monosodium (4-Amino-1-hydroxy-1-phosphonobutyl)phosphonate

A2456-25G A2456-5G
TCIAA2456-25GEA 0 CNY
TCIAA2456-25G TCIAA2456-5G
Alendronate sodium trihydrate ≥97.0% (by titrimetric analysis)
Alendronate sodium trihydrate
分子式: C₄H₁₂NaNO₇P₂·3H₂O
分子量: 325.12 g/mol
存储条件: Freezer
MDL编号: MFCD01748233
CAS号: 121268-17-5
Merck Index: 14,00229

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 97.0 %
Drying loss 16.1 to 17.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR