Amisulpride ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: 4-Amino-N-[(1-ethyl-2-pyrrolidinyl)methyl]-5-ethylsulphonyl-2-methoxybenzamide

A2450-1G A2450-100MG
TCIAA2450-1GEA 0 CNY
TCIAA2450-1G TCIAA2450-100MG
Amisulpride ≥98.0% (by HPLC, titration analysis)
Amisulpride
分子式: C₁₇H₂₇N₃O₄S
分子量: 369.48 g/mol
存储条件: Refrigerator
MDL编号: MFCD00866691
CAS号: 71675-85-9
EINECS: 275-831-7
Merck Index: 14,00485

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 125.0 to 129.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR